콘텐츠로 건너뛰기
Merck
  • CREB1 gene polymorphisms combined with environmental risk factors increase susceptibility to major depressive disorder (MDD).
CREB1 gene polymorphisms combined with environmental risk factors increase susceptibility to major depressive disorder (MDD).
International journal of clinical and experimental pathology (2015-03-11)
Peng Wang, Yanjie Yang, Xiuxian Yang, Xiaohui Qiu, Zhengxue Qiao, Lin Wang, Xiongzhao Zhu, Hong Sui, Jingsong Ma
초록
Major depressive disorder (MDD) is one of the most severe psychiatric disorders. The objective of this study was to explore the effects of CREB1 gene polymorphisms on risk of developing MDD and the joint effects of gene-environment interactions. Genotyping was performed by Taqman allelic discrimination assay among 586 patients and 586 healthy controls. A significant impact on rs6740584 genotype distribution was found for childhood trauma (P = 0.015). We did not find an association of CREB1 polymorphisms with MDD susceptibility. However, we found a significantly increased risk associated with the interactions of CREB1 polymorphisms and drinking (OR = 11.67, 95% CI = 2.52-54.18; OR = 11.52, 95% CI = 2.55-51.95 for rs11904814; OR = 4.18, 95% CI = 1.87-9.38; OR = 5.02, 95% CI = 2.27-11.14 for rs6740584; OR = 7.58, 95% CI = 2.05-27.98; OR = 7.59, 95% CI = 2.12-27.14 for rs2553206; OR = 8.37, 95% CI = 3.02-23.23; OR = 7.84, 95% CI = 2.93-20.98 for rs2551941). We also noted that CREB polymorphisms combined with family harmony and childhood trauma conferred increased susceptibility for MDD. In conclusion, polymorphisms in the CREB gene may not be independently associated with MDD risk, but they are likely to confer increased susceptibility by interacting with environmental risk factors in the Chinese population.