콘텐츠로 건너뛰기
Merck
  • RNA Interference Using c-Myc-Conjugated Nanoparticles Suppresses Breast and Colorectal Cancer Models.
RNA Interference Using c-Myc-Conjugated Nanoparticles Suppresses Breast and Colorectal Cancer Models.
Molecular cancer therapeutics (2015-02-20)
Naveen K Tangudu, Vinod K Verma, Tristan D Clemons, Syed S Beevi, Trevor Hay, Ganesh Mahidhara, Meera Raja, Rekha A Nair, Liza E Alexander, Anant B Patel, Jedy Jose, Nicole M Smith, Bogdan Zdyrko, Anne Bourdoncle, Igor Luzinov, K Swaminathan Iyer, Alan R Clarke, Lekha Dinesh Kumar
초록
In this article, we report the development and preclinical validation of combinatorial therapy for treatment of cancers using RNA interference (RNAi). RNAi technology is an attractive approach to silence genes responsible for disease onset and progression. Currently, the critical challenge facing the clinical success of RNAi technology is in the difficulty of delivery of RNAi inducers, due to low transfection efficiency, difficulties of integration into host DNA and unstable expression. Using the macromolecule polyglycidal methacrylate (PGMA) as a platform to graft multiple polyethyleneimine (PEI) chains, we demonstrate effective delivery of small oligos (anti-miRs and mimics) and larger DNAs (encoding shRNAs) in a wide variety of cancer cell lines by successful silencing/activation of their respective target genes. Furthermore, the effectiveness of this therapy was validated for in vivo tumor suppression using two transgenic mouse models; first, tumor growth arrest and increased animal survival was seen in mice bearing Brca2/p53-mutant mammary tumors following daily intratumoral treatment with nanoparticles conjugated to c-Myc shRNA. Second, oral delivery of the conjugate to an Apc-deficient crypt progenitor colon cancer model increased animal survival and returned intestinal tissue to a non-wnt-deregulated state. This study demonstrates, through careful design of nonviral nanoparticles and appropriate selection of therapeutic gene targets, that RNAi technology can be made an affordable and amenable therapy for cancer.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Myc
Sigma-Aldrich
MISSION® esiRNA, targeting human MYC