콘텐츠로 건너뛰기
Merck
  • Loss of methylation at GNAS exon A/B is associated with increased intrauterine growth.
Loss of methylation at GNAS exon A/B is associated with increased intrauterine growth.
The Journal of clinical endocrinology and metabolism (2015-01-21)
Anne-Claire Bréhin, Cindy Colson, Stéphanie Maupetit-Méhouas, Virginie Grybek, Nicolas Richard, Agnès Linglart, Marie-Laure Kottler, Harald Jüppner
초록
GNAS is one of few genetic loci that undergo allelic-specific methylation resulting in the parent-specific expression of at least four different transcripts. Due to monoallelic expression, heterozygous GNAS mutations affecting either paternally or maternally derived transcripts cause different forms of pseudohypoparathyroidism (PHP), including autosomal-dominant PHP type Ib (AD-PHP1B) associated with loss of methylation (LOM) at exon A/B alone or sporadic PHP1B (sporPHP1B) associated with broad GNAS methylation changes. Similar to effects other imprinted genes have on early development, we recently observed severe intrauterine growth retardation in newborns, later diagnosed with pseudopseudohypoparathyroidism (PPHP) because of paternal GNAS loss-of-function mutations. This study aimed to determine whether GNAS methylation abnormalities affect intrauterine growth. Birth parameters were collected of patients who later developed sporPHP1B or AD-PHP1B, and of their healthy siblings. Comparisons were made to newborns affected by PPHP or PHP1A. As newborns, AD-PHP1B patients were bigger than their healthy siblings and well above the reference average; increased sizes were particularly evident if the mothers were unaffected carriers of STX16 deletions. SporPHP1B newborns were slightly above average for weight and length, but their overgrowth was less pronounced than that of AD-PHP1B newborns from unaffected mothers. LOM at GNAS exon A/B due to maternal STX16 deletions and the resulting biallelic A/B expression are associated with enhanced fetal growth. These findings are distinctly different from those of PPHP patients with paternal GNAS exons 2-13 mutations, whose birth parameters are almost 4.5 z-scores below those of AD-PHP1B patients born to healthy mothers.