콘텐츠로 건너뛰기
Merck
  • Renal effects of the novel selective adenosine A1 receptor blocker SLV329 in experimental liver cirrhosis in rats.
Renal effects of the novel selective adenosine A1 receptor blocker SLV329 in experimental liver cirrhosis in rats.
PloS one (2011-03-23)
Berthold Hocher, Susi Heiden, Karoline von Websky, Ayman M Arafat, Jan Rahnenführer, Markus Alter, Philipp Kalk, Dieter Ziegler, Yvan Fischer, Thiemo Pfab
초록
Liver cirrhosis is often complicated by an impaired renal excretion of water and sodium. Diuretics tend to further deteriorate renal function. It is unknown whether chronic selective adenosine A(1) receptor blockade, via inhibition of the hepatorenal reflex and the tubuloglomerular feedback, might exert diuretic and natriuretic effects without a reduction of the glomerular filtration rate. In healthy animals intravenous treatment with the novel A(1) receptor antagonist SLV329 resulted in a strong dose-dependent diuretic (up to 3.4-fold) and natriuretic (up to 13.5-fold) effect without affecting creatinine clearance. Male Wistar rats with thioacetamide-induced liver cirrhosis received SLV329, vehicle or furosemide for 12 weeks. The creatinine clearance of cirrhotic animals decreased significantly (-36.5%, p<0.05), especially in those receiving furosemide (-41.9%, p<0.01). SLV329 was able to prevent this decline of creatinine clearance. Mortality was significantly lower in cirrhotic animals treated with SLV329 in comparison to animals treated with furosemide (17% vs. 54%, p<0.05). SLV329 did not relevantly influence the degree of liver fibrosis, kidney histology or expression of hepatic or renal adenosine receptors. In conclusion, chronic treatment with SLV329 prevented the decrease of creatinine clearance in a rat model of liver cirrhosis. Further studies will have to establish whether adenosine A(1) receptor antagonists are clinically beneficial at different stages of liver cirrhosis.
MATERIALS
제품 번호
브랜드
제품 설명
Sigma-Aldrich
SLV320, ≥98% (HPLC)