62895
Lysine Decarboxylase Salt Broth
suitable for microbiology
About This Item
shelf life
limited shelf life, expiry date on the label
composition
bromocresol purple, 0.01 g/L
cresol red, 0.005 g/L
glucose, 0.5 g/L
L-lysine, 10 g/L
meat extract, 5 g/L
meat peptone (peptic), 5 g/L
pyridoxal, 0.005 g/L
sodium chloride, 30 g/L
final pH
6.5±0.2 (37 °C)
application(s)
microbiology
Application
Recommended by the "Schweizerisches Lebensmittelbuch" 5th ed., chapter 56A.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service