A5476
Agitoxin-3
>98% (HPLC), recombinant, expressed in E. coli, solid
Synonym(s):
Gly-Val-Pro-Ile-Asn-Val-Pro-Cys-Thr-Gly-Ser-Pro-Gln-Cys-Ile-Lys-Pro-Cys-Lys-Asp-Ala-Gly-Met-Arg-Phe-Gly-Lys-Cys-Met-Asn-Arg-Lys-Cys-His-Cys-Thr-Pro-Lys
Sign Into View Organizational & Contract Pricing
Select a Size
All Photos(1)
Select a Size
Change View
About This Item
recombinant
expressed in E. coli
assay
>98% (HPLC)
form
solid
solubility
H2O: soluble
storage temp.
−20°C
Amino Acid Sequence
Gly-Val-Pro-Ile-Asn-Val-Pro-Cys-Thr-Gly-Ser-Pro-Gln-Cys-Ile-Lys-Pro-Cys-Lys-Asp-Ala-Gly-Met-Arg-Phe-Gly-Lys-Cys-Met-Asn-Arg-Lys-Cys-His-Cys-Thr-Pro-Lys
Biochem/physiol Actions
Potent Shaker-like voltage-gated K+ channels and their mammalian homologs blocker.
Other Notes
Toxin in Leiurus quinquestriatus hebraeus
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service