Skip to Content
Merck
All Photos(1)

Key Documents

A8852

Sigma-Aldrich

A-68930 hydrochloride

≥98% (HPLC), powder

Synonym(s):

cis-(±)-1-(Aminomethyl)-3,4-dihydro-3-phenyl-1H-2-benzopyran-5,6-diol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17NO3 · HCl
CAS Number:
Molecular Weight:
307.77
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

off-white to tan

solubility

H2O: soluble ~9 mg/mL

originator

Abbott

storage temp.

−20°C

SMILES string

Cl.NC[C@@H]1O[C@@H](Cc2c(O)c(O)ccc12)c3ccccc3

InChI

1S/C16H17NO3.ClH/c17-9-15-11-6-7-13(18)16(19)12(11)8-14(20-15)10-4-2-1-3-5-10;/h1-7,14-15,18-19H,8-9,17H2;1H/t14-,15-;/m0./s1

InChI key

PQPGUUQPTSMLKU-YYLIZZNMSA-N

Application

A-68930 hydrochloride has been used as a D1 receptor agonist to study its effect on striatal phosphorylation of cyclin-dependent kinase 5 (Cdk5) at Tyr15 in mice. It has also been used to study its effect on the cAMP accumulation in C-terminal mutant D1Rs in transfected HEK 293 cells.

Biochem/physiol Actions

A-68930 hydrochloride is a cell-membrane impermeable selective D1 dopamine receptor agonist.
A-68930 hydrochloride might possess antipsychotic properties.

Features and Benefits

This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Salmi et al.
Neuroreport, 11(6), 1269-1272 (2000-05-19)
The present results demonstrate sedative effects of the DA D1 receptor agonist A 68930 (0.9-15 micromol kg(-1), s.c.) on rat spontaneous locomotor activity in an open field. The effects were particularly strong, and dose-dependent, for the ambulatory activity in the
H.A. AI-Naser et al.
Behavioural pharmacology, 5(2), 210-218 (1994-04-01)
A-68930 [1R, 3S)-1-aminomethyl-5,6-dihydroxy-3-phenylisochroman HCI] is a novel, potent and selective dopamine D1 receptor agonist. Previous reports have shown that the selective D1 partial agonist, SK & F 38393, reduced food consumption in rats and mice. The aim of this study
J W Kebabian et al.
American journal of hypertension, 3(6 Pt 2), 40S-42S (1990-06-01)
A68930 (5,6-dihydroxy-3-phenyl-1-aminomethyl-isochroman) is a potent (EC50 = 2.5 nmol/L) partial agonist at the D-1 dopamine receptor. In contrast, A68930 is a much weaker agonist (EC50 = 3,920 nmol/L) at the D-2 dopamine receptor. The orientation of the 3-phenyl substituent in
Yan Guo et al.
PloS one, 6(12), e29204-e29204 (2011-12-30)
The dopamine D₁ receptor (D₁R), a G protein-coupled receptor, plays a critical role in regulating blood pressure through its actions on renal hemodynamics and epithelial ion transport, which are highly linked to its intracellular trafficking. In this study, we generated
R Isacson et al.
Neuroscience, 124(1), 33-42 (2004-02-13)
The behavioral and biochemical effects of the full dopamine D(1/5) receptor agonists, dihydrexidine and (1R,3S)-1-aminomethyl-5,6-dihydroxy-3-phenylisochroman HCl (A 68930), were examined in rats. Both A 68930 (0-4.6 mg kg(-1), s.c.) and dihydrexidine (0-8.0 mg kg(-1), s.c.) caused a dose-dependent suppression of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service