Skip to Content
Merck
All Photos(2)

Key Documents

HPA017346

Sigma-Aldrich

Anti-ADGRG6 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FLJ14937, Anti-GPR126

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

KRSLEDEPRLVLWALLVYNATNNTNLEGKIIQQKLLKNNESLDEGLRLHTVNVRQLGHCLAMEEPKGYYWPSIQPSEYVLPCPDKPGFSASRICFYNATNPLVTYWGPVD

General description

GPR126 (G protein-coupled receptor 126) is a memeber of the adhesion family of G-protein coupled receptors (GPCRs) consisting of C1r-C1s, Uegf and Bmp1 (CUB) and pentraxin (PTX)-like modules and a mucin-like spacer, and seven transmembrane domains. It is present on the cell surface of endothelial cells.

Immunogen

adhesion G protein-coupled receptor G6

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GPR126 (G protein-coupled receptor 126) plays a major role in cell-adhesion. It has been reported that GPR126 has potentiality in inflammation and coagulation. It functions as a secreted ligand in development. In humans, GPR126 plays an essential role in myelination of axons of peripheral nerves. Mutation in GPR126 causes arthrogryposis multiplex congenital.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70223

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetsuo Moriguchi et al.
Genes to cells : devoted to molecular & cellular mechanisms, 9(6), 549-560 (2004-06-11)
We have identified and characterized a novel member of the G protein-coupled receptor (GPCR) family, termed DREG. DREG belongs to the LNB-TM7 subfamily and possesses a long amino-terminus that contains a CUB domain, a PTX domain, a hormone binding domain
Gianina Ravenscroft et al.
American journal of human genetics, 96(6), 955-961 (2015-05-26)
Arthrogryposis multiplex congenita is defined by the presence of contractures across two or more major joints and results from reduced or absent fetal movement. Here, we present three consanguineous families affected by lethal arthrogryposis multiplex congenita. By whole-exome or targeted
Christian Stehlik et al.
FEBS letters, 569(1-3), 149-155 (2004-07-01)
Using a signal sequence trap for selection of differentially expressed secretory and membrane proteins, we identified a novel member of the adhesion family of G-protein coupled receptors (GPCRs), termed vascular inducible GPCR (VIGR). VIGR contains C1r-C1s, Uegf and Bmp1 (CUB)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service