Skip to Content
Merck
All Photos(1)

Key Documents

L1915

Sigma-Aldrich

Lq2

>98% (HPLC), recombinant, expressed in E. coli, lyophilized powder

Synonym(s):

ChTx2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

Pricing and availability is not currently available.

recombinant

expressed in E. coli

assay

>98% (HPLC)

form

lyophilized powder

mol wt

4359 Da

storage temp.

−20°C

Amino Acid Sequence

Gln-Phe-Thr-Gln-Glu-Ser-Cys-Thr-Ala-Ser-Asn-Gln-Cys-Trp-Ser-Ile-Cys-Lys-Arg-Leu-His-Asn-Thr-Asn-Arg-Gly-Lys-Cys-Met-Asn-Lys-Lys-Cys-Arg-Cys-Tyr-Ser

Biochem/physiol Actions

ROMK1 inward rectifier potassium channel blocker; also blocks single K(Ca) channels in planar lipid bilayer (rat muscle); Lq2 action is on the extracellular side of the pore.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z Lu et al.
Biochemistry, 36(23), 6936-6940 (1997-06-10)
We have purified a protein inhibitor of an inward-rectifier K+ channel, ROMK1, from the venom of the scorpion Leiurus quinquestriatus var. hebraeus. The inhibitor is Lq2, a previously discovered blocker of voltage- and Ca2+-activated K+ channels. Mutations were made on
K Lucchesi et al.
The Journal of membrane biology, 109(3), 269-281 (1989-08-01)
Two charybdotoxin peptides were purified from venom of the Israeli scorpion, Leiurus quinquestriatus hebraeus. Microsequencing of the most abundant toxin, ChTX-Lq1, revealed identity with the 37-residue peptide previously sequenced by Gimenez-Gallego et al. [Gimenez-Gallego, G., et al., Proc. Natl. Acad.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service