Skip to Content
Merck
All Photos(1)

Key Documents

P3488

Sigma-Aldrich

Protein Kinase G Iα from bovine lung

≥95% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

PKG Iα, cGK Iα, cyclic Guanosine Monophosphate Protein Kinase I α

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106305

Pricing and availability is not currently available.

assay

≥95% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

74 kDa (monomer)

shipped in

dry ice

storage temp.

−20°C

Physical form

Solution in 10 mM phosphate buffer, pH 6.8, 1 mM EDTA, 15 mM β-mercaptoethanol, 10 KIU/ml aprotinin and 50% glycerol.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U Walter et al.
The Journal of biological chemistry, 255(8), 3757-3762 (1980-04-25)
A guanosine 3':5'-monophosphate (cGMP)-dependent protein kinase was purified from bovine lung using 8-(6-aminohexylamino)-cAMP-Sepharose. The activity of the purified enzyme was highly dependent on cGMP using histone f2b as a substrate. The self-phosphorylation of the purified enzyme was strongly inhibited by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service