MilliporeSigma
LANEXO® Inventory Manager
The fast track to an ultra-efficient, audit-ready lab
Say goodbye to manual stock keeping. Our LANEXO® inventory manager does it all for you. Comprised of web and mobile apps plus RFID labels, LANEXO® lets you easily register, track and monitor lab consumables, giving you instant access to real-time inventory data – like SDS, owner, opening and expiry dates, location, usage and disposal information – anytime, anywhere. Discover how our intuitive software solution boosts efficiency, compliance and safety in regulated labs.
Already a customer? Login here.
  • More efficient inventory management
  • Compliant and audit ready
  • Greater safety with lower operational risk
  • Intuitive and easy to implement
Features
Real-time stock monitoring
Real-time stock monitoring
The LANEXO® app captures digital data from RFID labels on chemical bottles, providing real-time info on storage locations, stock usage and remaining levels.
Automated opening detection
Automated opening detection
Smart Seals detect when bottles are opened and automatically update their expiry dates. Our RFID labels can be applied to consumables from any vendor, as well as to in-house preparations. 
Safe handling of consumables
Safe handling of consumables
Instant access to electronic safety data sheets (SDS) ensures that you store and handle consumables safely. Storage incompatibility alerts on the mobile app prevent risks before they happen.
Compliant and audit ready
Compliant and audit ready
Secure data storage, time-stamped audit trails and traceable records ensure that your lab is audit ready and compliant with FDA 21 CFR Part 11 and EU GMP Annex 11.
Seamless integration to existing systems
Seamless integration to existing systems
Our cloud-based software as a service (SaaS) can be easily incorporated into existing workstreams via API integration to ELN, ERP and LIMS.
Intuitive and easy to implement
Intuitive and easy to implement
Our mobile and web apps are simple to set up and operate, using an intuitive, straightforward interface that lab personnel can learn in less than a day. 
User and approval management
User and approval management
Add new users easily activate or deactivate permissions according to your lab's setup, requirements, and approval processes.
Bulk inventory count with UHF
Bulk inventory count with UHF
With a UHF handheld reader plus the LANEXO® mobile app and RFID labels, you can count your entire inventory in one go – within seconds.
Related Product Resources
Connect with an Expert today
Request a live demonstration today to experience how LANEXO® inventory manager can pave the way for more efficient, reliable and compliant scientific discovery.
Fields indicated by an * are required.
Highlights
New LANEXO® System to Improve Scientists’ Productivity in the Lab
New LANEXO® System to Improve Scientists’ Productivity in the Lab
The LANEXO® Lab Inventory, Safety and Compliance Management System, a new digital laboratory informatics solution, is designed to drastically reduce time in labs and improve data quality and traceability.
LANEXO® System Earns Gold in 2021 German Design Awards
LANEXO® System Earns Gold in 2021 German Design Awards
The smart RFID technology based inventory management system received the highest distinction for Excellent Communications Design in the app category. The annual German Design Awards are the international premier prize of the German Design Council and are among the...
Fast-tracking Lab Digitalization
Fast-tracking Lab Digitalization
Digitalizing lab inventory processes has been shown to save time and resources, freeing up scientists to actually do science.
Time is Money
Time is Money
The Hidden Cost of Inefficient Laboratory Practices: overview of the main inventory management challenges in fast-paced life science laboratories and how they directly impact research outcomes, costs and regulatory compliance.