Skip to Content
MilliporeSigma
(11β,16α)-21-(Acetyloxy)-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

(11β,16α)-21-(Acetyloxy)-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione

(11β,16α)-21-(Acetyloxy)-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Acetyldesonide, Desonide 21-acetate
Empirical Formula (Hill Notation):
C26H34O7
CAS Number:
Molecular Weight:
458.54

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
API Family
Reference Material Type