MilliporeSigma
Hexadecyltrimethylammonium bromide ≥98%

Hexadecyltrimethylammonium bromide

Hexadecyltrimethylammonium bromide ≥98%
Synonyms:
CTAB, Cetrimonium bromide, Cetyltrimethylammonium bromide, Palmityltrimethylammonium bromide, Cetyltrimethylammonium Bromide, Molecular Biology Grade, Hexadecyltrimethylammonium bromide
Linear Formula:
C16H33N(CH3)3Br
CAS Number:
Molecular Weight:
364.45

Products

Shipping
Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique