Skip to Content
MilliporeSigma
4′-Isopropylacetophenone AldrichCPR

4′-Isopropylacetophenone

4′-Isopropylacetophenone AldrichCPR
Synonym(s):
4′-Isopropylacetophenone, p-Isopropylacetophenone
Empirical Formula (Hill Notation):
C11H14O
CAS Number:
Molecular Weight:
162.23

Products

Product Category
Brand
Feature
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Package Size
Special Grade