MilliporeSigma
4-Nitrophenol solution 10 mM

4-Nitrophenol solution

4-Nitrophenol solution 10 mM
Synonym(s):
p-Nitrophenol solution
Empirical Formula (Hill Notation):
C6H5NO3
CAS Number:
Molecular Weight:
139.11

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing