MilliporeSigma
G 418 disulfate salt suitable for plant cell culture, BioReagent

G 418 disulfate salt

G 418 disulfate salt suitable for plant cell culture, BioReagent
Synonyms:
Antibiotic G418
Empirical Formula (Hill Notation):
C20H40N4O10 · 2H2SO4
CAS Number:
Molecular Weight:
692.71

Products

Shipping
Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Activity Spectrum
Application
Mode of Action
Technique

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Showing 1-3 of 3