MilliporeSigma
Acetic acid 30% for analysis EMSURE® Reag. Ph Eur

Acetic acid 30%

Acetic acid 30% for analysis EMSURE® Reag. Ph Eur
Synonyms:
Acetic acid solution
Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05

Products

Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Showing 1-1 of 1
Compare
Product Number
Description
Pricing