
Aminoglutethimide impurity D

Synonym(s):
3,3′-(Azodi-4,1-phenylene)bis[3-ethyl-2,6-piperidinedione], 3,3′-[Diazenediylbis(4,1-phenylene)]bis(3-ethylpiperidine-2,6-dione), Azoglutethimide
Empirical Formula (Hill Notation):
C26H28N4O4
CAS Number:
Molecular Weight:
460.52
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing