Anti-CD9 antibody produced in rabbit
Synonym(s):
.MRP-1, CD9 antigen, Leukocyte antigen MIC3, Motility-related protein, p24, Anti-BA2, Anti-BTCC-1, Anti-DRAP-27, Anti-GIG2, Anti-Growth-inhibiting gene 2 protein, Anti-Leukocyte antigen MIC3, Anti-MRP-1, Anti-Motility related protein, Anti-TSPAN29, Anti-p24 antigen
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique