Anti-FGF4 antibody produced in rabbit
Synonym(s):
Anti-FGF-4, Anti-Fibroblast growth factor 4 precursor, Anti-HBGF-4, Anti-HST, Anti-HST-1, Anti-Heparin secretory-transforming protein, Anti-Transforming protein KS3, Anti-Fibroblast growth factor 4
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Application
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing