
Anti-MAP3K7 antibody produced in rabbit

Synonym(s):
Anti-Mitogen-activated protein kinase kinase kinase 7 antibody produced in rabbit, Anti-TGF-β-activated kinase 1 antibody produced in rabbit, Anti-Transforming growth factor-β-activated kinase 1 antibody produced in rabbit, M3K7, MAP3K7, Mitogen-activated protein kinase kinase kinase 7, TGF-β- activated kinase 1, Transforming growth factor-β-activated kinase 1
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing