
Aripiprazole EP Impurity C

Synonym(s):
7-[4-[4-(2-Chlorophenyl)piperazin-1-yl]butoxy]-3,4-dihydroquinolin-2(1H)-one, 3-Deschloroaripiprazole
Empirical Formula (Hill Notation):
C23H28ClN3O2
CAS Number:
Molecular Weight:
413.94
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing