MilliporeSigma
BIOshell A160 Peptide C18, 2.7 μm HPLC Column 2.7 μm particle size, L × I.D. 5 cm × 4.6 mm

BIOshell A160 Peptide C18, 2.7 μm HPLC Column

BIOshell A160 Peptide C18, 2.7 μm HPLC Column 2.7 μm particle size, L × I.D. 5 cm × 4.6 mm

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Analyte Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Package Size
Particle Size (µm)
Pore Size (Å)
Technique

Products

Showing 1-14 of 14
Compare
Product Number
Description
Pricing