MilliporeSigma
BIOshell™ A400 Protein C4, 3.4 μm HPLC Column 3.4 μm particle size, L × I.D. 5 cm × 4.6 mm

BIOshell A400 Protein C4, 3.4 μm HPLC Column

BIOshell™ A400 Protein C4, 3.4 μm HPLC Column 3.4 μm particle size, L × I.D. 5 cm × 4.6 mm

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Package Size
Particle Size (µm)
Pore Size (Å)
Technique

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Showing 1-11 of 11