Skip to Content
MilliporeSigma
CFTR(inh)-172 ≥98% (HPLC), powder

CFTR Inhibitor-172

CFTR(inh)-172 ≥98% (HPLC), powder
Synonym(s):
5-[(4-Carboxyphenyl)methylene]-2-thioxo-3-[(3-trifluoromethyl)phenyl-4-thiazolidinone, CFTR Inhibitor-172
Empirical Formula (Hill Notation):
C18H10F3NO3S2
CAS Number:
Molecular Weight:
409.40

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment