Skip to Content
MilliporeSigma
Cyclopamine phyproof® Reference Substance

Cyclopamine

Cyclopamine phyproof® Reference Substance
Synonym(s):
Cyclopamine, V. californicum, 11-Deoxyjervine
Empirical Formula (Hill Notation):
C27H41NO2
CAS Number:
Molecular Weight:
411.62

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade