
Dantrolene Related Compound C

Synonym(s):
5-(4-Nitrophenyl)furfural, 5-(4-Nitrophenyl)-2-furancarboxaldehyde
Empirical Formula (Hill Notation):
C11H7NO4
CAS Number:
Molecular Weight:
217.18
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing