
Estrone-2,3,4-13C3 solution

Empirical Formula (Hill Notation):
13C3C15H22O2
CAS Number:
Molecular Weight:
273.34
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Format
Quality Segment
Isotope
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2