Skip to Content
Merck
Frozen Human Normal CD19+ B Cells 10 Million Cells/per vial- HLA Typed

Frozen Human Normal CD19+ B Cells

Frozen Human Normal CD19+ B Cells 10 Million Cells/per vial- HLA Typed

Products

Brand
Biological Source
Manufacturer
Physical Form
Tissue
Quality Segment
Package Size
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing