
Gelatin Peptone for Biotechnology and Microbiology

Synonym(s):
Peptone from gelatin
CAS Number:
Products
Product Category
Brand
Biological Source
pH Value
Physical Form
Tissue
Chemical Composition
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing