Skip to Content
MilliporeSigma
γ-Undecalactone ≥98%, FCC, FG

γ-Undecalactone

γ-Undecalactone ≥98%, FCC, FG
Synonym(s):
Undecanoic γ-lactone, Aldehyde C14
Empirical Formula (Hill Notation):
C11H20O2
CAS Number:
Molecular Weight:
184.28
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Gene Alias
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing