
Hydroxypropyl Beta Cyclodextrin

Synonym(s):
(2-Hydroxypropyl)-β-cyclodextrin
CAS Number:
EC Number:
Products
Brand
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing