
Importazole

Synonym(s):
N-(1-Phenylethyl)-2-(pyrrolidin-1-yl)quinazolin-4-amine, Importazole
Empirical Formula (Hill Notation):
C20H22N4
CAS Number:
Molecular Weight:
318.42
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Special Grade
Showing 1-2 of 2