
Levamlodipine

Synonym(s):
(4S)-2-[(2-Aminoethoxy)methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-3,5-pyridinedicarboxylic acid 3-ethyl 5-methyl ester, (S)-(-)-Amlodipine
Empirical Formula (Hill Notation):
C20H25ClN2O5
CAS Number:
Molecular Weight:
408.88
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing