
Lithium (trimethylsilyl)acetylide solution

Synonym(s):
(Trimethylsilyl)ethynyllithium
Linear Formula:
(CH3)3SiC≡CLi
CAS Number:
Molecular Weight:
104.15
Products
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing