Skip to Content
MilliporeSigma
MES ULTROL® Grade, ≥99% dry basis (titration), zwitterionic buffer useful in pH range 5.5-6.5

MES

MES ULTROL® Grade, ≥99% dry basis (titration), zwitterionic buffer useful in pH range 5.5-6.5
Synonym(s):
2-(N-Morpholino)ethanesulfonic acid, 4-Morpholineethanesulfonic acid, MES, Free Acid, ULTROL® Grade
Empirical Formula (Hill Notation):
C6H13NO4S
CAS Number:
Molecular Weight:
195.24

Products

Shipping
Product Category
Brand
Color
Feature
Formula Weight
pH Value
Physical Form
pK Value
Purity
Research Area
Quality Segment
Application