Skip to Content
US
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium hydroxide solution
Products
Product Category
bases (6)
Brand
Supelco (7)
Physical Form
Product Line
Agency
Quality Segment
Reaction Type
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
1.09114
Potassium hydroxide solution,
c(KOH) = 0.5 mol/l (0.5 N), in ethanol, reag. Ph. Eur., ready-to-use volumetric solution for titration in non-aqueous media, Titripur
®
Expand
View Pricing
1.05544
Potassium hydroxide solution,
c(KOH)=0.1 mol/l (0,1 N) in 2-propanol, DIN ISO 6618, ready-to-use for titration in non-aqueous media, Titripur
®
Expand
View Pricing
1.09115
Potassium hydroxide solution,
c(KCl) = 0.1 mol/L (1 N) in ethanol, reag. USP, ready-to-use volumetric solution for titration in non-aqueous media, Titripur
®
Expand
View Pricing
1.11587
Potassium hydroxide solution,
c(KOH) = 0.1 mol/l (0.1 N) in methanol, ready-to-use volumetric solution for titration for non-aqueous media, Titripur
®
Expand
View Pricing
1.05545
Potassium hydroxide solution,
47%, for analysis EMSURE
®
Expand
View Pricing
1.09351
Potassium hydroxide solution,
c(KOH) = 0.5 mol/l (0.5 N) in methanol, ready-to-use volumetric solution for titration in non-aqueous media, Titripur
®
Expand
View Pricing
1.11787
Potassium hydroxide solution,
c(KOH) = 2.0 mol/l (2 N) in methanol, ready-to-use volumetric solution for titration in non-aqueous media, Titripur
®
Expand
View Pricing