
Racecadotril for peak identification

Synonym(s):
Racecadotril, (RS)-Benzyl N-[3-(acetylthio)-2-benzylpropanoyl]glycinate, (±)-Acetorphan, Acetorphan, Cadotril, Redotil, Tiorfan, Zedott
Empirical Formula (Hill Notation):
C21H23NO4S
CAS Number:
Molecular Weight:
385.48
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing