Skip to Content
MilliporeSigma
Sodium Hydroxide, Pellets

Sodium hydroxide pellets

Sodium Hydroxide, Pellets
Synonym(s):
Sodium hydroxide, ‘Caustic soda’
Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Agency
Quality Segment
Application
Technique
Showing 1-2 of 2