Skip to Content
MilliporeSigma
Trypsin inhibitor from chicken egg white Type III-O (free of ovoinhibitor)

Trypsin inhibitor from chicken egg white

Trypsin inhibitor from chicken egg white Type III-O (free of ovoinhibitor)
Synonym(s):
Ovomucoid
CAS Number:
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Physical Form
Tissue
Quality Segment