
Zirconium oxide

Synonym(s):
Zirconium(IV) oxide, Zirconia
Linear Formula:
ZrO2
CAS Number:
Molecular Weight:
123.22
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Particle Size (µm)
Showing 1-1 of 1
Compare
Product Number
Description
Pricing