- Transforming growth factor-beta1 down-regulation of major histocompatibility complex class I in thyrocytes: coordinate regulation of two separate elements by thyroid-specific as well as ubiquitous transcription factors.
Transforming growth factor-beta1 down-regulation of major histocompatibility complex class I in thyrocytes: coordinate regulation of two separate elements by thyroid-specific as well as ubiquitous transcription factors.
Transforming growth factor (TGF)-beta1-decreased major histocompatibility complex (MHC) class I gene expression in thyrocytes is transcriptional; it involves trans factors and cis elements important for hormone- as well as iodide-regulated thyroid growth and function. Thus, in rat FRTL-5 thyrocytes, TGF-beta1 regulates two elements within -203 bp of the transcription start site of the MHC class I 5'-flanking region: Enhancer A, -180 to -170 bp, and a downstream regulatory element (DRE), -127 to -90 bp, that contains a cAMP response element (CRE)-like sequence. TGF-beta1 reduces the interaction of a NF-kappaB p50/fra-2 heterodimer (MOD-1) with Enhancer A while increasing its interaction with a NF-kappaB p50/p65 heterodimer. Both reduced MOD-1 and increased p50/p65 suppresses class I expression. Decreased MOD-1 and increased p50/p65 have been separately associated with the ability of autoregulatory (high) concentrations of iodide to suppress thyrocyte growth and function, as well as MHC class I expression. TGF-beta1 has two effects on the downstream regulatory element (DRE). It increases DRE binding of a ubiquitously expressed Y-box protein, termed TSEP-1 (TSHR suppressor element binding protein-1) in rat thyroid cells; TSEP-1 has been shown separately to be an important suppressor of the TSH receptor (TSHR) in addition to MHC class I and class II expression. It also decreases the binding of a thyroid-specific trans factor, thyroid transcription factor-1 (TTF-1), to the DRE, reflecting the ability of TGF-beta1 to decrease TTF-1 RNA levels. TGF-beta1-decreased TTF-1 expression accounts in part for TGF-beta1-decreased thyroid growth and function, since decreased TTF-1 has been shown to decrease thyroglobulin, thyroperoxidase, sodium iodide symporter, and TSHR gene expression, coincident with decreased MHC class I. Finally, we show that TGF-beta1 increases c-jun RNA levels and induces the formation of new complexes involving c-jun, fra-2, ATF-1, and c-fos, which react with Enhancer A and the DRE. TGF-beta1 effects on c-jun may be a pivotal fulcrum in the hitherto unrecognized coordinate regulation of Enhancer A and the DRE.