MilliporeSigma
  • Urinary cyclic GMP after treatment of gynecological cancer. A prognostic marker of clinical outcome.
Urinary cyclic GMP after treatment of gynecological cancer. A prognostic marker of clinical outcome.
Anticancer research (2007-08-19)
Anne Orbo, Målfrid Hanevik, Ragnhild Jaeger, Saskia Van Heusden, Georg Sager
ABSTRACT
The search for biological markers to predict malignant disease and its recurrence, or to monitor the effectiveness of treatment is a continuous process in medicine. Several years ago, urinary excretion of cGMP in urine was found to be a sensitive predictor in the follow-up of ovarian cancer and of monitoring treatment of cancer of the uterine cervix. In the present study, 27 patients with gynecological cancer, including cancer of the uterine cervix (n=13), cancer of the uterine corpus (n=8) and cancer of the ovaries (n=6), were monitored for 10 years. Blood and urinary samples were taken before primary treatment (baseline sample) and three months thereafter (three-month sample). The serum levels of CEA, CA-125 and PIIINP and urine excretion of cGMP and cAMP were determined. Creatinine levels in serum and urine were employed to determine renal clearance. After 10 years' observation of women with cancer of the uterine cervix, seven patients showed no relapse and cGMP levels in baseline samples and three-month samples were 36.8+/-4.1 and 24.9+/-4.4 nmol cGMP/micromol creatinine (mean+/-SEM, p<0.01), respectively. The levels in patients (n=6) with relapse after 10 years' observation were 32.8+/-4.0 (baseline sample) and 43.5+/-4.2 (three-month sample) nmol cGMP/micromol creatinine (mean+/-SEM, p<0.02). Among the patients treated for cancer of the uterine corpus (n=9), none showed recurrent disease within the observation period of 10 years. The cGMP levels fell from 37.9+/-6.3 (baseline sample) to 22.3+/-2.3 (three-month sample) nmol cGMP/micromol creatinine (p<0.005). In the patients with ovarian cancer (n=6), 4 patients relapsed during the observation period of 10 years. In these women the cGMP levels increased from 34.5+/-2.7 (baseline sample) to 46.3+/-4.7 nmol cGMP/micromol creatinine whilst in both patients without relapse the levels decreased from 31.8 (range: 26.5-37.1) to 27.3 (range: 25.7-28.8) nmol cGMP/micromol creatinine, respectively. The changes in levels of cAMP, CEA, CA-125 and PIINP did not show statistically significant differences. Early changes in cGMP levels appear to predict long-term prognosis in gynecological cancers.
Product Description
Supelco
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 80 Coated Filter, with cassettes unassembled
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
Supelco
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Supelco
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
Supelco
ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate