MilliporeSigma
  • A semiautomated test for microsatellite instability and its significance for the prognosis of sporadic endometrial cancer in northern Norway.
A semiautomated test for microsatellite instability and its significance for the prognosis of sporadic endometrial cancer in northern Norway.
International journal of gynecological pathology : official journal of the International Society of Gynecological Pathologists (2002-01-10)
A Ørbo, K Eklo, M Kopp
ABSTRACT
Archival histologic material from 105 women (median age 62 years) treated for endometrial cancer was investigated for the replication error phenotype indicated by the observation of widespread microsatellite instability (MSI). Polymerase chain reaction (PCR) of DNA isolated from paraffin-embedded tissue was analyzed for MSI using six microsatellite loci with a fluorescent-based detection system. Flow cytometry and morphometric investigation were performed in the same material for each of the patients. Twenty percent (21 of 105) of screened endometrial cancers were found to have high MSI at two or more of the loci tested. The mean detection frequency per marker was highest in the dinucleotide repeat sequence, D2S123, and the mononucleotide repeat sequences amplified by Bat 25 and Bat 26. Death from endometrial cancer was not related to the occurrence of MSI (p=0.6). There was no significant association between MSI and FIGO stage (p=0.5), myometrial invasion depth (p=0.8), histological grade (p=0.3), or vessel invasion (p=0.5). There were, however, more MSI cases among the group of diploid cases compared with the aneuploid and tetraploid group. MSI is not a valuable prognosticator for survival of sporadic endometrial cancer, and diploid cases are significantly more often MSI positive than aneuploid and tetraploid cases.
Product Description
Supelco
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 80 Coated Filter, with cassettes unassembled
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
Supelco
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Supelco
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
Supelco
ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate