MilliporeSigma
  • Determinants of cognitive outcome in survivors of out-of-hospital cardiac arrest.
Determinants of cognitive outcome in survivors of out-of-hospital cardiac arrest.
Resuscitation (2014-09-03)
Marte Ørbo, Per M Aslaksen, Kristina Larsby, Lena Norli, Christoph Schäfer, Pål M Tande, Torgil R Vangberg, Audny Anke
ABSTRACT
To describe cognitive functioning with neuropsychological tests and examine predictors of cognitive outcome in adult survivors of out-of-hospital cardiac arrest (OHCA) of cardiac cause. The study was prospective and took place at the University hospital of North-Norway. Only patients eligible of neuropsychological assessment three months after OHCA were asked to participate. Cognitive test performance was compared to large samples of age-corrected normative data. General linear models were used to determine predictors of a cognitive composite score and performance on separate cognitive tests. The predictors assessed were coma duration, hypothermia treatment and time to restoration of spontaneous circulation. We aimed to control for demographic variables, medical comorbidity and affective symptoms. 45 survivors (4 women) completed the assessment. Neuropsychological tests of fine motor functioning, memory, attention and executive functions were significantly below normative means. Depending on the test, impairment ranged from 9 to 31%. For twenty-five survivors (56%), all cognitive tests were within the normal range. Shorter coma duration and induced hypothermia treatment were associated with favourable cognitive outcomes and explained 45% of the variability in the cognitive composite score. Coma duration was predictive across all cognitive tests, hypothermia treatment of specific tests of memory, attention and executive functioning. Cognitive outcome was normal in more than half of the survivors. Shorter coma duration and induced hypothermia were associated with favourable cognitive outcomes in the participating survivors three months after OHCA. Institutional protocol number: 2009/1395.
Product Description
Supelco
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Supelco
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 80 Coated Filter, with cassettes unassembled
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
Supelco
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
Supelco
ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate