Merck
Inhibition of protein kinase C by calphostin C is light-dependent.
Biochemical and biophysical research communications (1991-04-15)
R F Bruns, F D Miller, R L Merriman, J J Howbert, W F Heath, E Kobayashi, I Takahashi, T Tamaoki, H Nakano
ABSTRACT
Calphostin C, a secondary metabolite of the fungus Cladosporium cladosporioides, inhibits protein kinase C by competing at the binding site for diacylglycerol and phorbol esters. Calphostin C is a polycyclic hydrocarbon with strong absorbance in the visible and ultraviolet ranges. In characterizing the activity of this compound, we unexpectedly found that the inhibition of [3H]phorbol dibutyrate binding was dependent on exposure to light. Ordinary fluorescent light was sufficient for full activation. The inhibition of protein kinase C activity in cell-free systems and intact cells also required light. Light-dependent cytotoxicity was seen at concentrations about 5-fold higher than those inhibiting protein kinase C.
Sigma-Aldrich
Calphostin C from Cladosporium cladosporioides, ≥90% (HPLC), powder