MilliporeSigma
  • In vitro spectrum of activity of finafloxacin, a novel, pH-activated fluoroquinolone, under standard and acidic conditions.
In vitro spectrum of activity of finafloxacin, a novel, pH-activated fluoroquinolone, under standard and acidic conditions.
Antimicrobial agents and chemotherapy (2011-06-29)
Will Stubbings, Pamela Leow, Goh Chee Yong, Falicia Goh, Barbara Körber-Irrgang, Michael Kresken, Rainer Endermann, Harald Labischinski
ABSTRACT
Finafloxacin is a novel fluoroquinolone that exhibits enhanced antibacterial activity under acidic conditions. The aim of this study was to define the in vitro pH-activity relationship. Finafloxacin exhibited optimal antibacterial activity between pH 5.0 and 6.0 at which MICs were 4- to 8-fold lower than those determined at neutral pH. These observations were then confirmed against a larger collection of bacteria. These data suggest that finafloxacin could potentially offer a therapeutic advantage within acidic foci of infection.
Sigma-Aldrich
Finafloxacin, ≥95% (HPLC)