MilliporeSigma
  • Promoter-associated proteins of EPAS1 identified by enChIP-MS - A putative role of HDX as a negative regulator.
Promoter-associated proteins of EPAS1 identified by enChIP-MS - A putative role of HDX as a negative regulator.
Biochemical and biophysical research communications (2018-03-27)
Arash Hamidian, Marica Vaapil, Kristoffer von Stedingk, Toshitsugu Fujita, Camilla U Persson, Pontus Eriksson, Srinivas Veerla, Katleen De Preter, Frank Speleman, Hodaka Fujii, Sven Påhlman, Sofie Mohlin
ABSTRACT
Presence of perivascular neuroblastoma cells with high expression of hypoxia inducible factor (HIF)-2α correlates with distant metastasis and aggressive disease. Regulation of HIFs are traditionally considered to occur post-translationally, but we have recently shown that HIF-2α is unconventionally regulated also at the transcriptional level in neuroblastoma cells. Regulatory factors binding directly to EPAS1 (encoding HIF-2α) to promote transcription are yet to be defined. Here, we employ the novel CRISPR/Cas9-based engineered DNA-binding molecule-mediated chromatin immunoprecipitation (enChIP) - mass spectrometry (MS) methodology to, in an unbiased fashion, identify proteins that associate with the EPAS1 promoter under normoxic and hypoxic conditions. Our enChIP analysis resulted in 27 proteins binding to the EPAS1 promoter in neuroblastoma cells. In agreement with a general hypoxia-driven downregulation of gene transcription, the majority (24 out of 27) of proteins dissociate from the promoter at hypoxia. Among them were several nucleosome-associated proteins suggesting a general opening of chromatin as one explanation to induced EPAS1 transcription at hypoxia. Of particular interest from the list of released factors at hypoxia was the highly divergent homeobox (HDX) transcription factor, that we show inversely correlates with HIF-2α in neuroblastoma cells. We propose a putative model where HDX negatively regulates EPAS1 expression through a release-of-inhibition mechanism.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
L-Aspartic acid β-hydroxamate, serine racemase inhibitor