MilliporeSigma
  • A monoclonal antibody against the plant growth regulator, abscisic acid.
A monoclonal antibody against the plant growth regulator, abscisic acid.
Hybridoma (1994-12-01)
G M Banowetz, J R Hess, J G Carman
ABSTRACT
Monoclonal antibodies were prepared against the plant growth regulator abscisic acid (ABA) conjugated to keyhole limpet hemocyanin through C-4. One of these antibodies was characterized for use in a competition fluorescence enzyme-linked immunosorbent assay (F-ELISA). The antibody detected femtomole quantities of ABA when used in the F-ELISA and showed minimal cross-reactivity with ABA metabolites and structural analogs. Dilution analysis suggested that the F-ELISA could be used to determine the ABA content of methanolic extracts of crude samples of wheat seeds without further purification. The F-ELISA was used to determine the effect of seed priming on ABA levels in wheat seeds. The antibody also was used in a modified noncompetitive indirect ELISA to measure ABA content of wheat caryopses. The noncompetitive ELISA was more sensitive than the F-ELISA, although the F-ELISA had a broader measuring range. When our anti-ABA antibody and a commercially available anti-ABA antibody were compared by indirect ELISA, there were no significant differences between the ABA estimates.
Sigma-Aldrich
Abscisic Acid Immunoassay Detection Kit, Quantitate the levels of abscisic acid (ABA) in plant tissue