MilliporeSigma
Structural basis for dual-mode inhibition of the ABC transporter MsbA.
Nature (2018-05-04)
Hoangdung Ho, Anh Miu, Mary Kate Alexander, Natalie K Garcia, Angela Oh, Inna Zilberleyb, Mike Reichelt, Cary D Austin, Christine Tam, Stephanie Shriver, Huiyong Hu, Sharada S Labadie, Jun Liang, Lan Wang, Jian Wang, Yan Lu, Hans E Purkey, John Quinn, Yvonne Franke, Kevin Clark, Maureen H Beresini, Man-Wah Tan, Benjamin D Sellers, Till Maurer, Michael F T Koehler, Aaron T Wecksler, James R Kiefer, Vishal Verma, Yiming Xu, Mireille Nishiyama, Jian Payandeh, Christopher M Koth
ABSTRACT
The movement of core-lipopolysaccharide across the inner membrane of Gram-negative bacteria is catalysed by an essential ATP-binding cassette transporter, MsbA. Recent structures of MsbA and related transporters have provided insights into the molecular basis of active lipid transport; however, structural information about their pharmacological modulation remains limited. Here we report the 2.9 Å resolution structure of MsbA in complex with G907, a selective small-molecule antagonist with bactericidal activity, revealing an unprecedented mechanism of ABC transporter inhibition. G907 traps MsbA in an inward-facing, lipopolysaccharide-bound conformation by wedging into an architecturally conserved transmembrane pocket. A second allosteric mechanism of antagonism occurs through structural and functional uncoupling of the nucleotide-binding domains. This study establishes a framework for the selective modulation of ABC transporters and provides rational avenues for the design of new antibiotics and other therapeutics targeting this protein family.
Sigma-Aldrich
2,2-Dimethyl-1,3-propanediol, 99%
Sigma-Aldrich
Maltose solution, BioReagent, ~20% in H2O, Molecular Biology
Supelco
Discovery® Glycan SPE Tube, bed wt. 250 mg, volume 3 mL, pk of 54
Supelco
Discovery® Glycan SPE Tube, bed wt. 500 mg, volume 6 mL, pk of 30
Supelco
Discovery® Glycan SPE Tube, 5 g, volume 60 mL, pk of 16
Supelco
Discovery® Glycan SPE Bulk Packing, pkg of 50 g
Supelco
Discovery® Glycan SPE Tube, bed wt. 1 g, volume 12 mL, pk of 20